5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-carboxaldehyde
Supplier: Apollo Scientific
|
Warning
|
OR23279-1G
OR23279-5G
OR23279-10G
APOSOR23279-1GEA
132
EUR
APOSOR23279-1G
APOSOR23279-5G
APOSOR23279-10G
5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-carboxaldehyde
5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-carboxaldehyde
Formula:
C₉H₇NOS₂ MW: 209,288 g/mol Melting Pt: 130…131 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02682090 CAS Number: 499770-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...