Ethyl 4-(chlorosulphonyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate

Supplier: Apollo Scientific
Danger

Synonyms: 2,4-Dimethyl-5-(ethoxycarbonyl)-1H-pyrrole-3-sulphonyl chloride, 3-(Chlorosulphonyl)-2,4-dimethyl-5-(ethoxycarbonyl)-1H-pyrrole

OR23364-250MG OR23364-100MG
APOSOR23364-250MGEA 126 EUR
APOSOR23364-250MG APOSOR23364-100MG
Ethyl 4-(chlorosulphonyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate
Ethyl 4-(chlorosulphonyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate
Formula: C₉H₁₂ClNO₄S
MW: 265,714 g/mol
Melting Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD03407326
CAS Number: 368869-88-9
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR