Methyl 3-chloro-4-methylthiophene-2-carboxylate
Supplier: Apollo Scientific
OR24319-5G
APOSOR24319-5GEA
58
EUR
APOSOR24319-5G
Methyl 3-chloro-4-methylthiophene-2-carboxylate
Methyl 3-chloro-4-methylthiophene-2-carboxylate
Formula:
C₇H₇ClO₂S MW: 190,647 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00052072 CAS Number: 175137-11-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...