1-(3-Hydroxyprop-1-yl)piperidine 97%
Supplier: Apollo Scientific
Synonyms:
3-(Piperidin-1-yl)propan-1-ol, 1-Hydroxy-3-(piperidin-1-yl)propane
OR24928-10G
OR24928-25G
OR24928-5G
APOSOR24928-10GEA
58
EUR
APOSOR24928-10G
APOSOR24928-25G
APOSOR24928-5G
1-(3-Hydroxyprop-1-yl)piperidine 97%
1-(3-Hydroxyprop-1-yl)piperidine
Formula:
C₈H₁₇NO Boiling Pt: 94…95 °C (0,5 mm Hg) Density: 0,944 g/cm³ (25 °C) Flash Pt: 97,8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023781 CAS Number: 104-58-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...