Ethyl 2-[3-(2-amino-2-thioxoethyl)-4-oxo-3,4-dihydrophthalazin-1-yl]acetate

Supplier: Apollo Scientific

OR25555-1G OR25555-5G
APOSOR25555-1GEA 94.5 EUR
APOSOR25555-1G APOSOR25555-5G
Ethyl 2-[3-(2-amino-2-thioxoethyl)-4-oxo-3,4-dihydrophthalazin-1-yl]acetate
Ethyl 2-[3-(2-amino-2-thioxoethyl)-4-oxo-3,4-dihydrophthalazin-1-yl]acetate
Formula: C₁₄H₁₅N₃O₃S
MW: 305,352 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00174271
CAS Number: 131666-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR