Ethyl 1-(2-chloroacetyl)piperidine-4-carboxylate

Supplier: Apollo Scientific
Danger

OR25929-250MG OR25929-1G OR25929-5G
APOSOR25929-250MGEA 81.5 EUR
APOSOR25929-250MG APOSOR25929-1G APOSOR25929-5G
Ethyl 1-(2-chloroacetyl)piperidine-4-carboxylate
Ethyl 1-(2-chloroacetyl)piperidine-4-carboxylate
Formula: C₁₀H₁₆ClNO₃
MW: 233,692 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01480849
CAS Number: 318280-71-6
UN: 1760
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR