4-Chloro-1-methylquinolin-2(1H)-one

Supplier: Apollo Scientific
Warning

Synonyms: 4-Chloro-1,2-dihydro-1-methyl-2-oxoquinoline

OR26332-1G OR26332-250MG
APOSOR26332-1GEA 230 EUR
APOSOR26332-1G APOSOR26332-250MG
4-Chloro-1-methylquinolin-2(1H)-one
4-Chloro-1-methylquinolin-2(1H)-one
Formula: C₁₀H₈ClNO
MW: 193,63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00101322
CAS Number: 32262-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR