N-Hydroxybenzenecarboximidoyl cyanide 95%
Supplier: Apollo Scientific
OR27083-1G
OR27083-5G
APOSOR27083-1GEA
43.5
EUR
APOSOR27083-1G
APOSOR27083-5G
N-Hydroxybenzenecarboximidoyl cyanide 95%
N-Hydroxybenzenecarboximidoyl cyanide
Formula:
C₈H₆N₂O MW: 146,146 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00035803 CAS Number: 825-52-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...