5-(Methylthio)thiophene-2-carbonitrile
Supplier: Apollo Scientific
OR29197-10G
OR29197-1G
OR29197-5G
APOSOR29197-10GEA
691
EUR
APOSOR29197-10G
APOSOR29197-1G
APOSOR29197-5G
5-(Methylthio)thiophene-2-carbonitrile
5-(Methylthio)thiophene-2-carbonitrile
Formula:
C₆H₅NS₂ MW: 155,241 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00173741 CAS Number: 175205-78-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...