3-Chloro-4-hydroxybenzoic acid hemihydrate
Supplier: Apollo Scientific
|
Warning
|
OR2925-100G
OR2925-500G
OR2925-50G
APOSOR2925-100GEA
271
EUR
APOSOR2925-100G
APOSOR2925-500G
APOSOR2925-50G
3-Chloro-4-hydroxybenzoic acid hemihydrate
3-Chloro-4-hydroxybenzoic acid hemihydrate
Formula:
2C₇H₅ClO₃·H₂O Melting Pt: 168…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD02662353 CAS Number: 128161-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...