5-(4-Chlorophenyl)-2-methylfuran-3-carbonyl chloride

Supplier: Apollo Scientific
Danger

OR29472-1G OR29472-5G
APOSOR29472-1GEA 122 EUR
APOSOR29472-1G APOSOR29472-5G
5-(4-Chlorophenyl)-2-methylfuran-3-carbonyl chloride
5-(4-Chlorophenyl)-2-methylfuran-3-carbonyl chloride
Formula: C₁₂H₈Cl₂O₂
MW: 255,097 g/mol
Melting Pt: 125…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00275557
CAS Number: 175276-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR