(2,6-Dichloropyridin-4-yl)methyl [3-(2,6-dichlorophenyl)-5-methylisoxazol-4-yl]carbamate

Supplier: Apollo Scientific
Warning

Synonyms: (2,6-Dichloropyridin-4-yl)methyl [3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazol-4-yl]carbamate, 2,6-Dichloro-4-[({[3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazol-4-yl]carbamoyl}oxy)methyl]pyridine

OR29546-250MG OR29546-50MG
APOSOR29546-250MGEA 116 EUR
APOSOR29546-250MG APOSOR29546-50MG
(2,6-Dichloropyridin-4-yl)methyl [3-(2,6-dichlorophenyl)-5-methylisoxazol-4-yl]carbamate
(2,6-Dichloropyridin-4-yl)methyl [3-(2,6-dichlorophenyl)-5-methylisoxazol-4-yl]carbamate
Formula: C₁₇H₁₁Cl₄N₃O₃
MW: 447,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00207908
CAS Number: 261622-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR