2-Methyl-1,3-thiazole-4-carbonitrile
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Cyano-2-methyl-1,3-thiazole, 2-Methylthiazole-4-carbonitrile
OR29666-1G
OR29666-250MG
APOSOR29666-1GEA
74.2
EUR
APOSOR29666-1G
APOSOR29666-250MG
2-Methyl-1,3-thiazole-4-carbonitrile
2-Methyl-1,3-thiazole-4-carbonitrile
Formula:
C₅H₄N₂S MW: 124,164 g/mol Melting Pt: 58…61 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00068103 CAS Number: 21917-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...