{[({2-[(4-Chlorobenzyl)thio]pyrimidin-4-yl}methylene)amino]oxy}(2,6-dichloropyridin-4-yl)methanone

Supplier: Apollo Scientific
Warning

OR29720-250MG OR29720-50MG
APOSOR29720-250MGEA 116 EUR
APOSOR29720-250MG APOSOR29720-50MG
{[({2-[(4-Chlorobenzyl)thio]pyrimidin-4-yl}methylene)amino]oxy}(2,6-dichloropyridin-4-yl)methanone
{[({2-[(4-Chlorobenzyl)thio]pyrimidin-4-yl}methylene)amino]oxy}(2,6-dichloropyridin-4-yl)methanone
Formula: C₁₈H₁₁Cl₃N₄O₂S
Storage Temperature: Ambient
MDL Number: MFCD00126320
CAS Number: 263755-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR