3-Amino-5-bromoisoquinoline

Supplier: Apollo Scientific
Warning

OR300062-500MG OR300062-1G OR300062-250MG
APOSOR300062-500MGEA 545 EUR
APOSOR300062-500MG APOSOR300062-1G APOSOR300062-250MG
3-Amino-5-bromoisoquinoline
3-Amino-5-bromoisoquinoline
Formula: C₉H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD15526591
CAS Number: 1192815-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR