3-Bromo-6-chloroisoquinoline

Supplier: Apollo Scientific
Warning

OR300092-1G OR300092-250MG
APOSOR300092-1GEA 810 EUR
APOSOR300092-1G APOSOR300092-250MG
3-Bromo-6-chloroisoquinoline
3-Bromo-6-chloroisoquinoline
Formula: C₉H₅BrClN
Storage Temperature: Ambient
MDL Number: MFCD18793407
CAS Number: 1276056-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR