7-Chloroisoquinolin-1-ol
Supplier: Apollo Scientific
|
Warning
|
OR300118-1G
OR300118-250MG
APOSOR300118-1GEA
130
EUR
APOSOR300118-1G
APOSOR300118-250MG
7-Chloroisoquinolin-1-ol
7-Chloroisoquinolin-1-ol
Formula:
C₉H₆ClNO MW: 179,603 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08436984 CAS Number: 24188-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...