3-Chloro-6-methoxyisoquinoline

Supplier: Apollo Scientific
Warning

Synonyms: 3-Chloro-6-methoxy-2-azanaphthalene

OR300122-1G OR300122-50MG
APOSOR300122-1GEA 527 EUR
APOSOR300122-1G APOSOR300122-50MG
3-Chloro-6-methoxyisoquinoline
3-Chloro-6-methoxyisoquinoline
Formula: C₁₀H₈ClNO
MW: 193,63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044725
CAS Number: 82117-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR