3-Cyclopropylpyrazin-2-amine

Supplier: Apollo Scientific
Warning

OR300238-1G OR300238-250MG
APOSOR300238-1GEA 429 EUR
APOSOR300238-1G APOSOR300238-250MG
3-Cyclopropylpyrazin-2-amine
3-Cyclopropylpyrazin-2-amine
Formula: C₇H₉N₃
MW: 135.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19215706
CAS Number: 1190969-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR