3-Bromoquinoline-7-carboxylic acid

Supplier: Apollo Scientific
Warning

OR300369-1G OR300369-250MG OR300369-100MG
APOSOR300369-1GEA 1020 EUR
APOSOR300369-1G APOSOR300369-250MG APOSOR300369-100MG
3-Bromoquinoline-7-carboxylic acid
3-Bromoquinoline-7-carboxylic acid
Formula: C₁₀H₆BrNO₂
MW: 252,064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20327982
CAS Number: 1344046-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR