2-Chloro-5-iodonicotinonitrile
Supplier: Apollo Scientific
|
Warning
|
OR300387-1G
OR300387-250MG
APOSOR300387-1GEA
64
EUR
APOSOR300387-1G
APOSOR300387-250MG
2-Chloro-5-iodonicotinonitrile
2-Chloro-5-iodonicotinonitrile
Formula:
C₆H₂ClIN₂ MW: 264,451 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09909360 CAS Number: 766515-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...