(5-Bromo-3-chloropyridin-2-yl)methanamine hydrochloride 95%

Supplier: Apollo Scientific
Warning

OR300395-1G OR300395-250MG OR300395-100MG
APOSOR300395-1GEA 493 EUR
APOSOR300395-1G APOSOR300395-250MG APOSOR300395-100MG
(5-Bromo-3-chloropyridin-2-yl)methanamine hydrochloride 95%
(5-Bromo-3-chloropyridin-2-yl)methanamine hydrochloride
Formula: C₆H₆BrClN₂·ClH
MW: 257,943 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22689891
CAS Number: 1416714-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR