1-(Methylsulphonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%

Supplier: Apollo Scientific
Warning

OR300403-1G OR300403-250MG
APOSOR300403-1GEA 223 EUR
APOSOR300403-1G APOSOR300403-250MG
1-(Methylsulphonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
1-(Methylsulphonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formula: C₁₀H₁₇BN₂O₄S
MW: 272.129 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18383275
CAS Number: 944994-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR