6-Methylpyridine-2-carboxamidine hydrochloride 95+%
Supplier: Apollo Scientific
OR300668-1G
OR300668-5G
APOSOR300668-1GEA
219
EUR
APOSOR300668-1G
APOSOR300668-5G
6-Methylpyridine-2-carboxamidine hydrochloride 95+%
6-Methylpyridine-2-carboxamidine hydrochloride
Formula:
C₇H₉N₃·ClH MW: 171,627 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09952702 CAS Number: 112736-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...