Ethyl 1-benzylaziridine-2-carboxylate

Supplier: Apollo Scientific

Synonyms: 1-Benzylaziridine-2-carboxylic acid ethyl ester

OR300794-1G OR300794-250MG OR300794-500MG OR300794-2.5G OR300794-5G
APOSOR300794-1GEA 393 EUR
APOSOR300794-1G APOSOR300794-250MG APOSOR300794-500MG APOSOR300794-2.5G APOSOR300794-5G
Ethyl 1-benzylaziridine-2-carboxylate
Ethyl 1-benzylaziridine-2-carboxylate
Formula: C₁₂H₁₅NO₂
MW: 205,253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17011809
CAS Number: 34943-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR