4-Bromo-1-methyl-1H-1,2,3-triazole 98%
Supplier: Apollo Scientific
|
Warning
|
OR301020-1G
OR301020-5G
OR301020-250MG
APOSOR301020-1GEA
75.4
EUR
APOSOR301020-1G
APOSOR301020-5G
APOSOR301020-250MG
4-Bromo-1-methyl-1H-1,2,3-triazole 98%
4-Bromo-1-methyl-1H-1,2,3-triazole
Formula:
C₃H₄BrN₃ Melting Pt: 98…101 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD10696344 CAS Number: 13273-53-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...