Ethyl oxo(2-oxocyclopent-1-yl)acetate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl 2-oxo-2-(2-oxocyclopentyl)acetate, Ethyl (2-oxocyclopent-1-yl)glyoxylate, Ethyl 2-oxo-2-(2-oxocyclopent-1-yl)ethanoate
OR301038-1G
OR301038-5G
APOSOR301038-1GEA
14.7
EUR
APOSOR301038-1G
APOSOR301038-5G
Ethyl oxo(2-oxocyclopent-1-yl)acetate
Ethyl oxo(2-oxocyclopent-1-yl)acetate
Formula:
C₉H₁₂O₄ MW: 184,189 g/mol Boiling Pt: 129…130 °C (7 mm Hg) Melting Pt: 26…31 °C Density: 1,37 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00019311 CAS Number: 39163-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...