(2-Chloroethoxy)acetic acid 97%
Supplier: Apollo Scientific
|
Danger
|
OR301184-1G
OR301184-5G
APOSOR301184-1GEA
81.2
EUR
APOSOR301184-1G
APOSOR301184-5G
(2-Chloroethoxy)acetic acid 97%
(2-Chloroethoxy)acetic acid
Formula:
C₄H₇ClO₃ Boiling Pt: 130 °C (3,5 mm Hg) Melting Pt: 26…33 °C Density: 1,33 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD17014832 CAS Number: 14869-41-1 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...