4,6-Dichloropyrimidin-2-ol
Supplier: Apollo Scientific
Synonyms:
4,6-Dichloropyrimidin-2(1H)-one
OR301195-1G
OR301195-250MG
APOSOR301195-1GEA
463
EUR
APOSOR301195-1G
APOSOR301195-250MG
4,6-Dichloropyrimidin-2-ol
4,6-Dichloropyrimidin-2-ol
Formula:
C₄H₂Cl₂N₂O MW: 164,977 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14702755 CAS Number: 6297-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...