4-Bromo-5-nitro-1H-imidazole

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-5-nitroimidazole

OR301330-1G OR301330-5G
APOSOR301330-1GEA 75.7 EUR
APOSOR301330-1G APOSOR301330-5G
4-Bromo-5-nitro-1H-imidazole
4-Bromo-5-nitro-1H-imidazole
Formula: C₃H₂BrN₃O₂
MW: 191,971 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11501602
CAS Number: 6963-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR