3-Chloro-2-hydroxypropyl acetate 95%

Supplier: Apollo Scientific

OR302162-5G OR302162-1G
APOSOR302162-5GEA 300 EUR
APOSOR302162-5G APOSOR302162-1G
3-Chloro-2-hydroxypropyl acetate 95%
3-Chloro-2-hydroxypropyl acetate
Formula: C₅H₉ClO₃
MW: 152,576 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01698128
CAS Number: 24573-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR