(6-(Dimethylamino)-4-methylpyridin-3-yl)boronic acid 96%

Supplier: Apollo Scientific

Synonyms: 2-(Dimethylamino)-4-methylpyridine-5-boronic acid

OR302300-1G OR302300-500MG
APOSOR302300-1GEA 509 EUR
APOSOR302300-1G APOSOR302300-500MG
(6-(Dimethylamino)-4-methylpyridin-3-yl)boronic acid 96%
(6-(Dimethylamino)-4-methylpyridin-3-yl)boronic acid
Formula: C₈H₁₃BN₂O₂
MW: 180,012 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12912694
CAS Number: 535934-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR