(3aR,6aS)-tert-Butyl 5-oxohexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate 95+%

Supplier: Apollo Scientific
Warning

Synonyms: cis-tert-Butyl 5-oxohexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate

OR302643-10G OR302643-1G OR302643-5G
APOSOR302643-10GEA 297 EUR
APOSOR302643-10G APOSOR302643-1G APOSOR302643-5G
(3aR,6aS)-tert-Butyl 5-oxohexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate 95+%
(3aR,6aS)-tert-Butyl 5-oxohexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate
Formula: C₁₂H₁₉NO₃
MW: 225,284 g/mol
Melting Pt: 70…71 °C
Storage Temperature: Ambient
MDL Number: MFCD18782907
CAS Number: 146231-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR