1H-Imidazo[4,5-c]pyridine-2(3H)-thione
Supplier: Apollo Scientific
OR302737-1G
OR302737-5G
APOSOR302737-1GEA
78.3
EUR
APOSOR302737-1G
APOSOR302737-5G
1H-Imidazo[4,5-c]pyridine-2(3H)-thione
1H-Imidazo[4,5-c]pyridine-2(3H)-thione
Formula:
C₆H₅N₃S MW: 151.189 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09701424 CAS Number: 7239-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...