1,3-Bis(bromomethyl)-5-bromobenzene 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
α,α',5-Tribromo-m-xylene
OR303130-1G
OR303130-500MG
OR303130-250MG
APOSOR303130-1GEA
338
EUR
APOSOR303130-1G
APOSOR303130-500MG
APOSOR303130-250MG
1,3-Bis(bromomethyl)-5-bromobenzene 95%
1,3-Bis(bromomethyl)-5-bromobenzene
Formula:
C₈H₇Br₃ MW: 342,853 g/mol Melting Pt: 95…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00192715 CAS Number: 51760-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...