Methyl 6-(4-methylpiperazin-1-yl)pyridine-3-carboxylate

Supplier: Apollo Scientific

OR303142-1G OR303142-500MG
APOSOR303142-1GEA 164 EUR
APOSOR303142-1G APOSOR303142-500MG
Methyl 6-(4-methylpiperazin-1-yl)pyridine-3-carboxylate
Methyl 6-(4-methylpiperazin-1-yl)pyridine-3-carboxylate
Formula: C₁₂H₁₇N₃O₂
Melting Pt: 76…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00894965
CAS Number: 132144-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR