1,2-Dimethylpyridin-1-ium iodide
Supplier: Apollo Scientific
OR303150-1G
OR303150-500MG
OR303150-5G
APOSOR303150-1GEA
180
EUR
APOSOR303150-1G
APOSOR303150-500MG
APOSOR303150-5G
1,2-Dimethylpyridin-1-ium iodide
1,2-Dimethylpyridin-1-ium iodide
Formula:
C₇H₁₀N·I Storage Temperature: Ambient |
MDL Number:
MFCD00031867 CAS Number: 872-73-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...