4-(Naphthalen-2-yl)butanoic acid
Supplier: Apollo Scientific
Synonyms:
2-Naphthalenebutyricacid
OR303161-1G
OR303161-500MG
APOSOR303161-1GEA
538
EUR
APOSOR303161-1G
APOSOR303161-500MG
4-(Naphthalen-2-yl)butanoic acid
4-(Naphthalen-2-yl)butanoic acid
Formula:
C₁₄H₁₄O₂ MW: 214,26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00114681 CAS Number: 782-28-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...