2-Methyl-2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]propanenitrile
Supplier: Apollo Scientific
OR303187-1G
OR303187-500MG
APOSOR303187-1GEA
538
EUR
APOSOR303187-1G
APOSOR303187-500MG
2-Methyl-2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]propanenitrile
2-Methyl-2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]propanenitrile
Formula:
C₁₅H₂₁BN₂O₂ MW: 272,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22689176 CAS Number: 1402390-59-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...