Ethyl 6-{[(tert-butoxy)carbonyl]amino}pyridine-3-carboxylate

Supplier: Apollo Scientific
Warning

OR303203-1G OR303203-500MG OR303203-5G
APOSOR303203-1GEA 270 EUR
APOSOR303203-1G APOSOR303203-500MG APOSOR303203-5G
Ethyl 6-{[(tert-butoxy)carbonyl]amino}pyridine-3-carboxylate
Ethyl 6-{[(tert-butoxy)carbonyl]amino}pyridine-3-carboxylate
Formula: C₁₃H₁₈N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD23103501
CAS Number: 169280-82-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR