4-Cyano-1H-pyrrole-2-carboxylic acid 97+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Carboxy-4-cyano-1H-pyrrole, 5-Carboxy-1H-pyrrole-3-carbonitrile
OR303210-1G
OR303210-500MG
APOSOR303210-1GEA
336
EUR
APOSOR303210-1G
APOSOR303210-500MG
4-Cyano-1H-pyrrole-2-carboxylic acid 97+%
4-Cyano-1H-pyrrole-2-carboxylic acid
Formula:
C₆H₄N₂O₂ MW: 136.108 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09991904 CAS Number: 80242-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...