3,4-Dihydro-2H-1,4-benzoxazine-6-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
OR303219-500MG
APOSOR303219-500MGEA
661
EUR
APOSOR303219-500MG
3,4-Dihydro-2H-1,4-benzoxazine-6-carboxylic acid
3,4-Dihydro-2H-1,4-benzoxazine-6-carboxylic acid
Formula:
C₉H₉NO₃ MW: 179,173 g/mol Melting Pt: 163…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD11848179 CAS Number: 918789-44-3 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...