6-Amino-3H,4H-pyrido[2,3-b]pyrazin-3-one
Supplier: Apollo Scientific
OR303233-1G
OR303233-500MG
APOSOR303233-1GEA
671
EUR
APOSOR303233-1G
APOSOR303233-500MG
6-Amino-3H,4H-pyrido[2,3-b]pyrazin-3-one
6-Amino-3H,4H-pyrido[2,3-b]pyrazin-3-one
Formula:
C₇H₆N₄O MW: 162,149 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20697305 CAS Number: 874493-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...