5-Amino-3-methyl-1,2-thiazole-4-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
OR303238-1G
OR303238-500MG
APOSOR303238-1GEA
270
EUR
APOSOR303238-1G
APOSOR303238-500MG
5-Amino-3-methyl-1,2-thiazole-4-carboxylic acid
5-Amino-3-methyl-1,2-thiazole-4-carboxylic acid
Formula:
C₅H₆N₂O₂S MW: 158,178 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06376749 CAS Number: 22131-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...