Methyl 2-(cyclohexylamino)-5-nitrobenzoate
Supplier: Apollo Scientific
OR303242-1G
OR303242-500MG
APOSOR303242-1GEA
180
EUR
APOSOR303242-1G
APOSOR303242-500MG
Methyl 2-(cyclohexylamino)-5-nitrobenzoate
Methyl 2-(cyclohexylamino)-5-nitrobenzoate
Formula:
C₁₄H₁₈N₂O₄ MW: 278.304 g/mol Melting Pt: 132…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD23989316 CAS Number: 83909-56-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...