Dimesna

Supplier: Apollo Scientific
Warning

OR303346-1G OR303346-5G OR303346-250MG
APOSOR303346-1GEA 74.1 EUR
APOSOR303346-1G APOSOR303346-5G APOSOR303346-250MG
Dimesna
Dimesna
Formula: C₄H₈O₆S₄·2Na
Melting Pt: >274 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00868218
CAS Number: 16208-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR