3-Bromo-9-methyl-9H-carbazole
Supplier: Apollo Scientific
|
Warning
|
OR303352-1G
OR303352-500MG
OR303352-5G
APOSOR303352-1GEA
48
EUR
APOSOR303352-1G
APOSOR303352-500MG
APOSOR303352-5G
3-Bromo-9-methyl-9H-carbazole
3-Bromo-9-methyl-9H-carbazole
Formula:
C₁₃H₁₀BrN Melting Pt: 76…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00276495 CAS Number: 91828-08-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...