1-Aminoazetidin-3-ol dihydrochloride
Supplier: Apollo Scientific
OR304325-1G
OR304325-5G
APOSOR304325-1GEA
589
EUR
APOSOR304325-1G
APOSOR304325-5G
1-Aminoazetidin-3-ol dihydrochloride
1-Aminoazetidin-3-ol dihydrochloride
Formula:
C₃H₈N₂O·2ClH Storage Temperature: Ambient |
MDL Number:
MFCD30471612 CAS Number: 2007919-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...