(2S)-1-Methylazetidine-2-carboxylic acid

Supplier: Apollo Scientific

OR304329-1G OR304329-250MG OR304329-500MG
APOSOR304329-1GEA 2050 EUR
APOSOR304329-1G APOSOR304329-250MG APOSOR304329-500MG
(2S)-1-Methylazetidine-2-carboxylic acid
(2S)-1-Methylazetidine-2-carboxylic acid
Formula: C₇H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD30146430
CAS Number: 1860067-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR