2-Bromo-1-cyclobutylethanone

Supplier: Apollo Scientific

OR304358-10G OR304358-1G OR304358-5G
APOSOR304358-10GEA 1090 EUR
APOSOR304358-10G APOSOR304358-1G APOSOR304358-5G
2-Bromo-1-cyclobutylethanone
2-Bromo-1-cyclobutylethanone
Formula: C₆H₉BrO
MW: 177.039 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847541
CAS Number: 128312-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR